Learn More
ABclonal Technology TRAP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2591920UL
Encompass_Preferred
This gene encodes a mitochondrial chaperone protein that is member of the heat shock protein 90 (HSP90) family The encoded protein has ATPase activity and interacts with tumor necrosis factor type I This protein may function in regulating cellular stress responses Alternate splicing results in multiple transcript variants
Catalog No. 50-000-12140