Learn More
Med Vet International Trapezoid Shaped Gel Positioner 10" L X 7" W X 3, J1282E, Each
Supplier:  Med Vet International JOR1282E
Encompass_Preferred
Gel Positioners are designed to aid in positioning patients for either surgery or radiographs. They provide firm but comfortable support. - Made of viscoelastic polymer so it has a soft tissue-like feel - Provides constant support and will not bottom out - Free from latex, silicon, and plasticizer - Will not leak or flow - Radiolucent - Reusable - Easy to clean with standard disinfectants - Compatible with warming blankets - 2 year warranty Trapezoid shaped gel positioner. Trapezoid. 10"L x 7"W x 3 1/2"H Note: Gel positioners are not warranted against damage caused by chewing or nail tears.
Catalog No. 50-005-81294