Learn More
Selleck Chemical LLC Trastuzumab-A2007-1MG-5
SDP
Supplier:  Selleck Chemical LLC A20071MG 5PC
Encompass_Preferred
Trastuzumab (anti-human HER2) is a humanized recombinant monoclonal antibody that binds to the extracellular domain of HER2 MW 145 53 KD
Catalog No. 50-313-8344