Learn More
Selleck Chemical LLC Travoprost S3738-2mg
SDP
Supplier:  Selleck Chemical LLC S37382MG
Encompass_Preferred
Travoprost (Travoprostum) is a PGF2a analog that was launched as an ophthalmic solution administered topically for the treatment of elevated intraocular hypertension as a result of open-angle glaucoma a common optic neuropathy and a leading cause of blindness It can act as an agonist of prostaglandin F receptor
Catalog No. 50-310-8376