Learn More
Bellco Glass, Inc. Tray,25x25cm
SDP
Supplier:  Bellco Glass, Inc. 774001010
Encompass_Preferred
Standard tray with pegs, *NOT* a stacking tray. Trays fit 7740 and 7743 series rocker & 7777 series 12v rocker. The "peg and grommet" design permits rapid platform changing.. MADE IN THE USA SIZE: 25x25cm
Catalog No. 50-194-6104