Learn More
Bellco Glass, Inc. Tray w/Posts,25X25cm
SDP
Supplier:  Bellco Glass, Inc. 774000010
Encompass_Preferred
Painted aluminum stackable tray for use with 7740-01010 platform tray and 7740/7743 rocker platforms.. MADE IN THE USA SIZE: 25x25cm
Catalog No. 50-194-6100