Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0478100MG
Encompass_Preferred
Trazodone hydrochloride is a triazolopyridine derivative that belongs to the class of serotonin receptor antagonists and reuptake inhibitors (SARIs). It has antidepressant and anti-insomnious activity, exerting antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors with minimal anticholinergic effects.
  • Triazolopyridine derivative
  • Serotonin receptor antagonist and reuptake inhibitor
  • Exhibits antidepressant activity
  • Possesses anti-insomnious activity
  • Antagonizes α1- and α2-adrenergic receptors
  • Antagonizes histamine H1 receptors
  • Shows minimal anticholinergic effects
  • Demonstrates neuroprotective effects
Catalog No. 50-004-06717