Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0478R5MG
Encompass_Preferred
Trazodone hydrochloride is an antidepressant classified as a serotonin receptor antagonist and reuptake inhibitor. It is primarily intended for research and analytical applications, particularly in the study of anxiety disorders, and serves as a reference standard.
  • Analytical standard for research
  • Used in analytical applications
  • Functions as a serotonin receptor antagonist and reuptake inhibitor
  • Employed for treatment of anxiety disorders
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Compatible with HPLC, GC, and MS techniques
Catalog No. 50-004-07813