Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | MFCD00079603 | 99.9% | 408.32 | C19H23Cl2N5O | 10 MG

Supplier: Medchemexpress LLC HYB0478R10MG
Trazodone hydrochloride is supplied as a high-purity analytical standard for research and analytical applications. It is provided as a solid reference material suitable for calibration, method development, and quality-control testing, packaged in small weighed amounts for laboratory use.
- High purity: 99.90%.
- Cas number: 25332-39-2.
- Molecular formula: C19H23Cl2N5O; molecular weight: 408.32.
- Available in small weighed sizes, including 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.
- Intended for research and analytical standard applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.