Learn More
SPEX CERTIPREP LLC 3P Chlorine Check Snip- Amperometric/Iodometric Titration 0.100ppm, 25/pk
Supplier:  SPEX CERTIPREP LLC 3P QCI148
Encompass_Preferred
(NSI by Zeptometrix) Designed for use with amperometric and/or colorimetric analytical techniques. Easier than glass ampule reference materials. To use, snip open the tube and quantitatively transfer the tube contents to a flask and bring to 200mL with reagent water. Yields a 0.100ppm working standard. No pipets needed. 25 tubes per pack.
Catalog No. NC1104956