Learn More
XenoTech,LLC Pooled B-naphthoflavone-treated SD rat liver microsomes, male, 0.5mL at 20mg/mL
SDP
Supplier:  XenoTech,LLC R1083
Encompass_Preferred
Pooled B-naphthoflavone-treated SD rat liver microsomes, male, 0.5mL at 20mg/mL
Catalog No. 50-722-700