Learn More
Waters Corp Chromatography Column, CEL2, 2.5 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186007655
Encompass_Preferred
The Trefoil CEL2 Column offers efficient chiral separations of enantiomers, stereoisomers, metabolites, degradants, and contaminants. It reduces method development time and enhances SuperCritical Fluid Chromatography (SFC) system performance, especially with mass spectrometry.
  • Optimized for ACQUITY UPC2 System.
  • Compatible with mass spectrometers.
  • Modified polysaccharide-coated stationary phase for broad chiral selectivity.
  • Reduces method development time.
  • Optimum peak shape and efficiencies.
  • Facilitates fast, easy separation of enantiomers, stereoisomers, metabolites, degradants, and contaminants.
  • Packing materials from ISO-certified facilities.
  • Individually tested; eCord Intelligent Chip provides performance chromatogram and certificate of batch analysis.
  • Tolerates operating pressures up to 6000 psi (415 Bar) and 40 °C.
Catalog No. 50-239-1989