Learn More
Selleck Chemical LLC Treosulfan
SDP
Supplier:  Selleck Chemical LLC S69585MG
Encompass_Preferred
Treosulfan (NSC 39069 Treosulphan) is an alkylating agent used for conventional and high-dose chemotherapy regimens Treosulfan is potently cytotoxic against pancreatic cancer cell lines
Catalog No. 50-000-02243