Learn More
ATCC Treponema denticola
Supplier:  ATCC 35405
Encompass
Treponema denticola strain a [CIP 103919, DSM 14222] is a whole-genome sequenced type strain that was isolated in Montreal from a human periodontal pocket. This bacterial strain produces methyl-accepting chemotaxis protein, major surface protein, and prolyl aminopeptidase.
Catalog No. 50-238-2364