Learn More
ATCC Treponema phagedenis; Kazan 8
Supplier:  ATCC 27087
Encompass
Treponema phagedenis strain Kazan 8 was isolated from a human with syphilis. This strain will not grow on a solid medium.
Catalog No. 50-238-3505