Learn More
Medchemexpress LLC Trequinsin hydrochloride | 78416-81-6 | 99.9% | 441.95 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY18740A100MG
Encompass_Preferred
Trequinsin hydrochloride is an extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE), with an IC50 of 0.25 nM. It is also an extremely potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin, and epinephrine.
  • Extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE).
  • Potent inhibitor of human platelet aggregation induced by ADP, collagen, thrombin, and epinephrine.
  • Cardiovascular and antihypertensive qualities.
  • Very potent antiplatelet activities.
  • Efficacious agonist of [Ca2+]i.
Catalog No. 50-003-88197