Learn More
Medchemexpress LLC Trequinsin hydrochloride | 78416-81-6 | 99.9% | 441.95 | C24H28ClN3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18740A10MG
Encompass_Preferred
Trequinsin hydrochloride is a research-grade phosphodiesterase (PDE) inhibitor supplied as the hydrochloride salt in solid form. It potently inhibits platelet cAMP PDE and reduces platelet aggregation in vitro, making it suitable for biochemical and cellular assays requiring a highly potent PDE modulator.
  • High purity (99.9%).
  • Potent PDE inhibition (IC50 ≈ 0.25 nM).
  • Soluble in DMSO and water; may require ultrasonic assistance.
  • Stable when stored sealed at 4°C; in solution store at -80°C for long-term.
  • Supplied in small solid aliquots suitable for assay preparation.
Catalog No. 50-002-98467