Learn More
Medchemexpress LLC HY-13543 5mg Medchemexpress, Tretazicar CAS:21919-05-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135435MG
Encompass_Preferred
Medchemexpress, HY-13543 5mg Tretazicar CAS:21919-05-1 Tretazicar (CB 1954), an antitumor prodrug, is highly selective against the Walker 256 rat tumour line. Tretazicar is enzymatically activated to generate a bifunctional agent, which can form DNA-DNA interstrand cross-links. Tretazicar in rat cells involves the reduction of its 4-nitro group to a 4-hydroxylamine by the enzyme NAD(P)H:quinone oxidoreductase 1 (NQO1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9878