Learn More
Selleck Chemical LLC Tretinoin (Retinoic acid) 50mg 302-79-4 All-trans Retinoic Acid,ATRA
SDP
Supplier:  Selleck Chemical LLC S165350MG
Encompass_Preferred
Tretinoin (Retinoic acid, All-trans Retinoic Acid, ATRA), which is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR), can induce granulocytic differentiation and apoptosis in acute promyelocytic leukemia (APL) cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1296630