Learn More
Medchemexpress LLC HY-125006 10mg , TRi-1 CAS:246020-68-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12500610MG
Encompass_Preferred
HY-125006 10mg TRi-1 CAS: 246020-68-8 TRi-1 is a potent, specific and irreversible inhibitor of cytosolic thioredoxin reductase 1 (TXNRD1), with an IC50 of 12 nM. TRi-1 has little mitochondrial toxicity for anticancer therapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0002