Learn More
Medchemexpress LLC Tri(2-methylphenyl)phosphine | 6163-58-2 | 99.99% | C21H21P | 500 G
SDP
Supplier:  Medchemexpress LLC HY70026500G
Encompass_Preferred
Tri(2-methylphenyl)phosphine is an organic phosphorus ligand, a type of chemical compound used in various chemical reactions. It is primarily utilized as a drug intermediate, meaning it serves as a building block in the synthesis of more complex pharmaceutical compounds. This ligand has the ability to form complexes with metal salts, which is a crucial characteristic for its application in catalysis and other chemical processes.
  • Acts as an organic phosphorus ligand
  • Used as a precursor in the synthesis of new chemical entities
  • Capable of forming complexes with metal salts
Catalog No. 50-004-34304