Learn More
Medchemexpress LLC Tri(Amino-PEG3-amide)-amine | 2523025-40-1 | 98.0% | C33H69N7O12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14024950MG
Encompass_Preferred
Tri(Amino-PEG3-amide)-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • For research use only.
Catalog No. 50-003-57217