Learn More
Hoefer, Inc 1.0ml Tri-coded Tube, 96-format, Rack - Sterilization. Pack contains 930 each.
SDP
Supplier:  Hoefer, Inc ST101001R
Encompass_Preferred
1.0ml Tri-coded Tube, 96-format, Sterilization, 96/rack, 10racks/case. Features and Benefits:Manufactured in clean workshop complying with GMP 8 and certified with ISO 9001 and13485.Strict quality control, Tri-Coded system, providing readability and uniqueness, and unequaled sample audit traceability.Laser-etched code on the tube bottom and side, easy to be read, scratch resistant.Super-toughened materials with standing temperatures from -196℃ to 120℃ for various storage condition.Same material for cap and tube, effectively preventing the uneven expansion during freeze-thaw cycles.Non-silicone seal with unique thread to prevent over-tightening.RNase / DNase and pyrogen free.Direct code-reading with the tubes on rack with SBS format.
Catalog No. NC2956958