Learn More
Medchemexpress LLC Tri-GalNAc-COOH | 1953146-81-0 | ≥99.0% | C75H134N10O35 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13948210MG
Encompass_Preferred
tri-GalNAc-COOH is a trivalent N-acetylgalactosamine (GalNAc) ligand that binds the asialoglycoprotein receptor (ASGPR). Supplied as a carboxyl-functionalized building block, it is intended for lysosome-targeting chimera (LYTAC) research and hepatocyte-directed delivery studies. The compound is available as solid material and as a 10 mM solution in DMSO, and is reported with high purity and a molecular weight of approximately 1,736 g·mol-1.
  • Trivalent GalNAc ligand for ASGPR targeting.
  • Carboxyl-functionalized handle suitable for conjugation.
  • Designed for LYTAC and hepatocyte delivery research applications.
  • Offered in multiple solid sizes and a DMSO solution format.
  • High reported purity to support reproducible experiments.
  • Recommended storage at -80°C under inert atmosphere for stability.
Catalog No. 50-002-60801