Learn More
Medchemexpress LLC Tri(GalNAc-PEG2)-PEG4-cyclo(Ac-DCAW-{Arg(3F-Ph)-PEG2}-LGELVWCT) (Cys2-Cys12) | 99.5% | 3488.76 | C152H236FN25O62S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17191010MG
Encompass_Preferred
tri(GalNAc-PEG2)-PEG4-cyclo(Ac-DCAW-{Arg(3F-Ph)-PEG2}-LGELVWCT) (Cys2-Cys12) is an asialoglycoprotein receptor (ASGPR) ligand supplied as a purified solid for research use. It is intended for lysosome targeting chimera (LYTAC) research and should be handled by qualified personnel in appropriately equipped laboratories. Store at -20°C protected from light and under inert gas; in solution store at -80°C (≤6 months) or -20°C (≤1 month).
  • Asialoglycoprotein receptor (ASGPR) ligand for LYTAC research.
  • High purity: 99.5%.
  • Molecular weight 3488.76; formula C152H236FN25O62S2.
  • Supplied as a solid; available in 500 μg-100 mg sizes.
  • Documentation: datasheet, COA, SDS, HNMR, LCMS, RP-HPLC available.
  • For research use only; not for human use.
Catalog No. 50-002-03572
Only null left
Add to Cart
Request a Quote