Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tri(GalNAc-PEG2)-PEG4-cyclo(Ac-DCAW-Arg(3F-Ph)-PEG2-LGELVWCT) (Cys2-Cys12) | 99.5% | 3488.76 | C152H236FN25O62S2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1719105MG

Encompass_Preferred

This synthetic peptide-GalNAc conjugate is an asialoglycoprotein receptor (ASGPR) ligand intended for lysosome targeting chimera (LYTAC) research. It combines tri-N-acetylgalactosamine units with PEG spacers and a cyclic peptide core to enable ASGPR-mediated uptake of conjugated cargos.

  • Is an ASGPR ligand suitable for lysosome targeting chimera research.
  • Peptide-based conjugate bearing tri-N-acetylgalactosamine (tri-GalNAc) moieties linked through PEG spacers.
  • Molecular weight 3,488.76.
  • Purity 99.46% by HPLC.
  • Appearance white to off-white solid; hygroscopic in solution.
  • Soluble in DMSO at 33.33 mg/mL (9.55 mM); ultrasonic agitation recommended.
  • Store at -20°C protected from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-04073


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.