Learn More
Strem, An Ascensus Company CAS# 688-73-3. 25g. Tri-n-butyltin hydride, min. 95%. MFCD00009416
SDP
Supplier:  Strem, An Ascensus Company 93508125G
Encompass
CAS# 688-73-3. 25g. Tri-n-butyltin hydride, min. 95%. MFCD00009416. Molecular Weight: 291.04. Molecular Formula: (C4H9)3SnH. Color/form: colorless liq. Strem# 93-5081. http://www.strem.com/catalog/v/93-5081/
Catalog No. 50-901-15630