Learn More
Medchemexpress LLC Tri-N-tridecyl 3-(ethyl(methyl)amino)propanoate | 1566559-80-5 | C57H114N4O6 | 5 MG

Supplier: Medchemexpress LLC HY1531375MG
Tri-N-tridecyl 3-(ethyl(methyl)amino)propanoate is a biodegradable lipid prepared by the conjugate addition of alkylamines to acrylates. This compound can be utilized in various drug delivery systems for the delivery of polynucleotides, such as siRNA.
- Purity: 98.71%
- Molecular weight: 951.54
- Appearance: Liquid, colorless to light yellow
- Density: 0.943±0.06 g/cm³
- Shipping: Room temperature in continental US, may vary elsewhere
- Storage: -20°C, protect from light, stored under nitrogen
- Storage in solvent: -80°C for 6 months, -20°C for 1 month (protect from light, stored under nitrogen)
- Solubility in vitro: Ethanol at ≥ 100 mg/mL (105.09 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.