Learn More
Andwin Scientific TRI-POUR BEAKER 50ML 100/PKTR
SDP
Supplier:  Andwin Scientific 60968
Encompass_Preferred
The Tri-Corn Beakers feature three dripless pouring spouts. This product is resistant to most chemicals and is autoclavable. It can resist temperatures up to 121C. The beakers have volume capacity of 50 mL and has graduations at 5 mL. This is sold in a pack of 100.
Catalog No. 50-996-322