Learn More
Medchemexpress LLC Triacetonamine (2,2,6,6-tetramethyl-4-piperidone) | 826-36-8 | MFCD00005975 | 155.24 | 1000 G
SDP
Supplier:  Medchemexpress LLC HYN11311000G
Encompass_Preferred
Triacetonamine is utilized as an intermediate in the synthesis of pharmaceutical products, pesticides, and photostabilizers for polymers. It exhibits oral activity and can induce acute liver failure (ALF) in rats.
  • Used as an intermediate for the synthesis of pharmaceutical products, pesticides, and photostabilizers for polymers
  • Has oral activity
  • Can induce acute liver failure (ALF) in rats
Catalog No. 50-004-15958