Learn More
Abcam Triacsin C, acyl-CoA synthetase inhibitor, 1MG
Supplier:  Abcam AB1418881MG
Encompass_Preferred
MW 207.27 Da, Purity >90%. Potent, specific acyl-CoA synthetase inhibitor (Ki values are 15 nM and 2 μM for Faa2p and Faa4p respectively). Inhibits synthesis of cholesteryl ester and triacylglycerol in vitro. Vasodilator agent. Orally active. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3339