Learn More
Cayman Chemical TrIadImefon 100mg
SDP
Supplier:  Cayman Chemical 18714100 MG
Encompass_Preferred
A triazole fungicide that inhibits lanosterol 14α-demethylase, interfering with ergosterol biosynthesis, and blocks gibberellin biosynthesis
Catalog No. 50-297-9392