Learn More
Medchemexpress LLC Triadimefon-d4 | 99.5% | 297.77 | C14H12D4ClN3O2 | 1mg
SDP
Supplier:  Medchemexpress LLC HYW7061801MG
Encompass_Preferred
( )-Triadimefon-d4 is deuterium labeled Triadimefon Triadimefon is a triazole fungicide used to control powdery mildew rusts and other fungal pests on grains fruit and vegetable crops turf shrubs and trees Triadimefon inhibits lanosterol 14 -demethylase interfering with oxidative demethylation reactions in the ergosterol biosynthesis pathway of fungi and also blocks gibberellin biosynthesis[1]
Catalog No. 50-002-70473