Learn More
Medchemexpress LLC Triadimenol-d4 | 2121989-56-6 | 500 UG
Supplier: Medchemexpress LLC HYB0851S500UG
Triadimenol-d4 is the deuterium labeled Triadimenol, a triazole fungicide widely used in agriculture. This compound, while effective as a fungicide, exhibits certain toxicity to animals. Its deuterium labeling, using stable heavy isotopes, allows it to be used as a tracer for quantitation during drug development. Deuteration has been shown to potentially impact the pharmacokinetic and metabolic profiles of drugs.
- Deuterium labeled Triadimenol
- Functions as a triazole fungicide
- Used in agriculture
- Exhibits certain toxicity to animals
- Incorporates stable heavy isotopes as tracers for drug development quantitation
- May affect pharmacokinetic and metabolic profiles of drugs