Learn More
Medchemexpress LLC Triamcinolone | 124-94-7 | 99.8% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB03281G
Encompass_Preferred
Triamcinolone is a long-acting corticosteroid with anti-inflammatory, anti-oedematous, anti-proliferative, anti-angiogenetic, immunomodulatory, and neuroprotective effects. It works by binding to glucocorticoid receptors.
  • Alleviates various dermatitis, immune diseases, and ocular diseases.
  • Intended for research and analytical applications.
  • Target: Glucocorticoid receptor, interleukin related.
  • Research areas include: cardiovascular disease, inflammation/immunology, endocrinology, and cancer.
Catalog No. 50-004-06364