Learn More
Medchemexpress LLC Triamcinolone | 124-94-7 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB03285G
Encompass_Preferred
Triamcinolone is a long-acting corticosteroid that exhibits anti-inflammatory, anti-oedematous, anti-proliferative, anti-angiogenetic, immunomodulatory, and neuroprotective effects by binding to glucocorticoid receptors. It is used to alleviate several dermatitis, immune diseases, and ocular diseases.
  • Long-acting corticosteroid.
  • Exhibits anti-inflammatory, anti-oedematous, anti-proliferative, anti-angiogenetic, immunomodulatory, and neuroprotective effects.
  • Binds to glucocorticoid receptors.
  • Used to alleviate dermatitis, immune diseases, and ocular diseases.
  • Demonstrates dose-dependent inhibition of bovine retinal endothelial cell proliferation.
  • Lowers neovascular cell count in retinopathy of prematurity animal models.
  • Investigated in numerous clinical trials.
Catalog No. 50-004-06375