Learn More
Medchemexpress LLC Triamcinolone acetonide | 76-25-5 | 99.95% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB063610MM/1ML
Encompass_Preferred
Triamcinolone acetonide is a synthetic corticosteroid known for its anti-inflammatory properties. It inhibits basic fibroblast growth factor (bFGF) induced proliferation of retinal endothelial cells and can be used in the study of diseases such as atopic dermatitis. It also activates macrophages with anti-inflammatory characteristics.
  • Reduces chondrocyte viability and leads to cartilage destruction.
  • Activates macrophages with anti-inflammatory characteristics.
  • Inhibits bFGF-induced proliferation of retinal endothelial cells.
Catalog No. 50-004-08255