Learn More
Medchemexpress LLC Triamcinolone acetonide | 76-25-5 | 99.95% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0636R50MG
Encompass_Preferred
Triamcinolone acetonide is a compound that inhibits basic fibroblast growth factor (bFGF) induced proliferation of retinal endothelial cells. It also reduces chondrocyte viability and activates macrophages with anti-inflammatory characteristics, making it suitable for studying diseases like atopic dermatitis.
  • Inhibits basic fibroblast growth factor (bFGF) induced proliferation of retinal endothelial cells
  • Reduces chondrocyte viability
  • Activates macrophages with anti-inflammatory characteristics
  • Can be used in the study of diseases such as atopic dermatitis
Catalog No. 50-004-07697