Learn More
Medchemexpress LLC Triamcinolone acetonide-d6 | 352431-33-5 | 99.1% | 440.53 | C24H25D6FO6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0636S310MG
Encompass_Preferred
Triamcinolone acetonide-d6 is the deuterium-labeled analogue of triamcinolone acetonide intended for use as a tracer or internal standard in quantitative analytical methods. It is supplied in small analytical pack sizes for laboratory research use.
  • Deuterium-labeled internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
  • Useful as a tracer in pharmacokinetic and metabolic studies.
  • High reported purity (about 99.1%) for reliable analytical performance.
  • Provided in small pack sizes suitable for analytical workflows.
Catalog No. 50-002-98121