Learn More
Medchemexpress LLC Triamcinolone hexacetonide | 5611-51-8 | 532.64 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYU0010310MG
Encompass_Preferred
Triamcinolone hexacetonide is a long-acting steroid commonly used for subacute and chronic inflammatory joint diseases. It is intended for research use only.
  • Produces a marked, dose-dependent protective effect in chemically induced articular cartilage damage models.
  • Reduces fibrillation and osteophytes in guinea pigs.
  • Reduces chondrocyte cloning, cell loss, osteophyte formation, and fibrillation in rabbits.
  • Local application can lead to reduced mechanical sensitivity and enhanced regeneration in lingual nerve injury.
  • Reformulated iso-osmolar Triamcinolone hexacetonide shows no evidence of deleterious effects on retina function or structure.
Catalog No. 50-004-49624