Learn More
Medchemexpress LLC Triamcinolone hexacetonide | 5611-51-8 | 532.64 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYU0010350MG
Encompass_Preferred
Triamcinolone hexacetonide is a long-acting steroid commonly used in the treatment of subacute and chronic inflammatory joint diseases. This compound has demonstrated a protective effect against articular cartilage damage and can aid in nerve regeneration.
  • Long-acting steroid for inflammatory conditions
  • Protective effect against articular cartilage damage
  • Reduces fibrillation, osteophytes, and cell loss
  • Supports nerve regeneration and reduces mechanical sensitivity
Catalog No. 50-004-49625