Learn More
Medchemexpress LLC Triamcinolone (Standard) | 124-94-7 | 98.02% | 50 MG

Supplier: Medchemexpress LLC HYB0328R50MG
Triamcinolone (Standard) is an analytical standard and reference material intended for research and analytical applications. It is utilized in qualitative, quantitative, and methodological research experiments.
- Used in techniques such as HPLC, GC, and MS
- Structure classification: Steroid
- Initial source: Plants (Solanaceae, Solanum lyratum Thunberg)
- Appearance: Solid
- Color: White to off-white
- Molecular weight: 394.43
- Formula: C₂₁H₂₇FO₆
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.