Learn More
Apexbio Technology LLC Triamterene 396-01-0 100mg
SDP
Supplier:  Apexbio Technology LLC B2275100
Encompass_Preferred
Triamterene (CAS 396-01-0) is a small molecule inhibitor that targets epithelial sodium channels (ENaC) It acts through a voltage-dependent mechanism to impede sodium transport across epithelial membranes In electrophysiological assays triamterene demonstrates an inhibitory concentration (IC50) of 4 5 M for ENaC blockade This compound is utilized in research to investigate sodium channel physiology ion transport processes in epithelial tissues and the pharmacological modulation of renal sodium handling
Catalog No. 50-224-0609