Learn More
Medchemexpress LLC Triamterene-d5 | 1189922-23-3 | MFCD09841356 | 99.8% | 258.29 | C12H6D5N7 | 10 MG

Supplier: Medchemexpress LLC HYB0575S10MG
Triamterene-d5 is a deuterium-labeled triamterene analog supplied as a high-purity analytical standard for use in mass spectrometry, tracer studies, and quantitative assays. It offers consistent isotopic labeling and stability for reliable internal standard performance.
- Deuterium-labeled (d5) for use as an internal standard in MS assays.
- High purity (99.8%) for accurate quantitation.
- Molecular weight 258.29 g/mol for precise mass calculations.
- Available in small milligram pack sizes for analytical workflows.
- Stable when stored as recommended: powder -20°C; in solvent -80°C short term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.