Learn More
Selleck Chemical LLC Triapine 10mg 200933-27-3 3-AP
SDP
Supplier:  Selleck Chemical LLC S747010MG
Encompass_Preferred
Triapine (3-AP) is a potent ribonucleotide reductase (RNR) inhibitor with broad spectrum antitumor activity by inhibiting DNA synthesis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4825