Learn More
YMC America, Inc TRIART C8 120A 5UM 75X4.6MM
SDP
Supplier:  YMC America, Inc TO12S05L546WT
Encompass_Preferred
TO12S05-L546WT: Triart C8, 120A, 5um, 75x4.6mm Endcapped Column
Catalog No. 50-394-88