Learn More
Medchemexpress LLC Triazabicyclodecene | 5807-14-7 | 99.8% | C7H13N3 | 10 G
SDP
Supplier:  Medchemexpress LLC HY5915610G
Encompass_Preferred
Triazabicyclodecene is a bicyclic glycone compound that serves as a drug intermediate, used in the formation of amides or esters. This product is intended for research use only and is an off-white to light yellow solid.
  • Can be used to form amides or esters
  • High purity of 99.8%
  • Available in various solid quantities and as a 10 mM solution
  • Detailed solubility information provided for in vitro and in vivo applications
  • Comprehensive documentation available
Catalog No. 50-004-34000