Learn More
Cayman Chemical TrIazavIrIn 10mg
SDP
Supplier:  Cayman Chemical 3605210 MG
Encompass_Preferred
An antiviral agent; inhibits lung viral accumulation in a mouse model of H5N1 influenza A infection at 1 mg/kg; inhibits platelet aggregation in a rat model of LPS-induced hypercytokinemia at 20 mg/kg
Catalog No. 50-291-2778