Learn More
Medchemexpress LLC Triazolopyridinone (3-hydroxytriazolo[4,3-a]pyridine) | 6969-71-7 | MFCD00022632 | 99.6% | 135.12 g/mol | C6H5N3O | 50 G

Supplier: Medchemexpress LLC HYW05358750G
Triazolopyridinone (3-hydroxytriazolo[4,3-a]pyridine) is a small-molecule intermediate used in organic synthesis, notably in the synthesis of trazodone and related triazolopyridine derivatives. It is supplied as a high-purity research reagent for synthetic and medicinal chemistry applications.
- Used as an intermediate in the synthesis of trazodone and related compounds.
- Synonym: 3-hydroxytriazolo[4,3-a]pyridine.
- CAS number 6969-71-7.
- Molecular formula C6H5N3O and molecular weight 135.12 g/mol.
- Offered in multiple package sizes up to 100 g.
- High reported purity (listed values around 99.6-100.0%).
- Stable at room temperature with documented shelf stability in the certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.