Learn More
Medchemexpress LLC Tribenuron-methyl | 101200-48-0 | 98.0% | 395.39 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111912
Encompass_Preferred
Tribenuron-methyl is a sulfonylurea herbicide agent that can also be used as a fungicide agent. It plays an important role in controlling weeds and diseases in wheat fields. This product is for research use only and not sold to patients.
  • Sulfonylurea herbicide agent.
  • Can be used as a fungicide agent.
  • Plays an important role in controlling weeds and diseases in wheat fields.
Catalog No. 50-004-39325